missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPEP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532724
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Gibbon, Monkey (93%) Panda, Rabbit, Horse (86%).
The protein encoded by this gene is a kidney membrane enzyme involved in the metabolism of glutathione and other similar proteins by dipeptide hydrolysis. The encoded protein is known to regulate leukotriene activity by catalyzing the conversion of leukotriene D4 to leukotriene E4. This protein uses zinc as a cofactor and acts as a disulfide-linked homodimer. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| DPEP1 | |
| Polyclonal | |
| Unconjugated | |
| DPEP1 | |
| AI327012; Dehydropeptidase-I; dipeptidase 1; dipeptidase 1 (renal); Dpep1; hRDP; MBD; Mbd1; MBD-1; MDP; membrane bound dipeptidase type II; membrane-bound dipeptidase 1; Microsomal dipeptidase; RDP; Renal dipeptidase; testicular tissue protein Li 57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P16444 | |
| DPEP1 | |
| Synthetic 14 amino acid peptide from internal region of human DPEP1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction