missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPEP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£173.00 - £238.00
Specifications
| Antigen | DPEP1 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DPEP1 Polyclonal antibody specifically detects DPEP1 in Human samples. It is validated for ELISASpecifications
| DPEP1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology | |
| 0.2 um filtered solution in PBS | |
| 1800 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| dehydropeptidase-I, dipeptidase 1, dipeptidase 1 (renal), EC 3.4.13.19, hRDP, MDPMBD1, RDPMicrosomal dipeptidase, Renal dipeptidase | |
| Produced in rabbits immunized with purified, recombinant Human DPEP1 (Accession#: P16444; Met1-Ser385) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title