missing translation for 'onlineSavingsMsg'
Learn More

DPD 1 green reagent solution, Lovibond™

Product Code. 10394643
This item is not returnable. View return policy
Product Code. 10394643 Supplier Lovibond Tintometer Supplier No. 471026

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DPD 1 green reagent solution

DPD 1 green reagent solution 6x15mL dropper bottle for approximatley 900 tests 0.02-4mg/L chlorine.

DPD 1 green reagent solution 6x15mL dropper bottle for approximatley 900 tests 0.02-4mg/L chlorine. 

Specifications

Chemical Name or Material Buffer Solution
compliance-icons compliance-icons
Product Identifier
  • DPD 1 reagent solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: sulfuric acid, N, N-Diethyl-1, 4-benzoldiamin-sulfat (1:1)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.