missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ DPD 1 Buffer Solution

Product Code. 13503890
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 13503890

Brand: Lovibond™ 471011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lovibond™ DPD 1 Buffer Solution is a liquid reagent for easy drop application.

  • Precise liquid preparation
  • Easy dropwise application
  • Should be stored tightly sealed, and away from sunlight and high temperatures
  • Shelf life: Two years from production date, provided that the bottles are stored within the temperature range +6°C to +10°C

Specifications

For Use With (Application) Determination of free chlorine
For Use With (Equipment) Photometer
Quantity 100 mL
Type DPD1 Buffer solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.