missing translation for 'onlineSavingsMsg'
Learn More

DPCR-1 Antibody (986124) [Allophycocyanin], Novus Biologicals™

Product Code. 30734239 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734239 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734239 Supplier Novus Biologicals Supplier No. FAB33471A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DPCR-1 Monoclonal antibody specifically detects DPCR-1 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPCR-1
Applications Immunocytochemistry
Classification Monoclonal
Clone 986124
Conjugate APC
Formulation PBS
Gene Alias bCX105N19.6, C6orf37, diffuse panbronchiolitis critical region 1, MGC126710, MGC126712, PBLTdiffuse panbronchiolitis critical region 1 protein
Host Species Mouse
Immunogen Human DPCR-1 synthetic peptide, Accession # Q3MIW9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135656
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.