missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPC AJ1951, xTFA, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methylalanyl-L-valyl-2-methylalanyl-L-alpha-glutamyl-L-isoleucyl-L-glutaminyl-L-leucyl-L-norleucyl-L-histidyl-L-glutaminyl-L-arginyl-L-alanyl-L-lysyl-L-tyrosinamide TFA salt |
| Molecular Formula | C76 H127 N23 O19 . x[C2 H F3 O2] |
| Quantity | 1 mg |
| Synonym | DPC AJ1951, xTFA |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?