missing translation for 'onlineSavingsMsg'
Learn More

Melford DPBS 1X Solution

Product Code. 30018230
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 30018230

Brand: Melford D221501000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phosphate Buffered Saline is a buffer solution commonly used in biological research.

Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.

Ready to use sterile 1X solution.

Ingredients:

Sodium Chloride: 137 mM

Potassium Chloride: 2.7 mM

Phosphate Buffer: 11.9 mM

pH: 7.4

Does not contain Calcium or Magnesium

Specifications

Quantity 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.