missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA581067
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DP2/TFDP2.
The gene is a member of the transcription factor DP family. The encoded protein forms heterodimers with the E2F transcription factors resulting in transcriptional activation of cell cycle regulated genes. Alternative splicing results in multiple transcript variants.
Specifications
| DP2 | |
| Polyclonal | |
| PBS with no preservative | |
| Q14188 | |
| TFDP2 | |
| A synthetic peptide corresponding to the C-terminus of the Human DP2/TFDP2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| TFDP2 | |
| BTPS2, DP2, DP2.5, DP3, GS | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 7029 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction