missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568914
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.
The gene is a member of the transcription factor DP family. The encoded protein forms heterodimers with the E2F transcription factors resulting in transcriptional activation of cell cycle regulated genes. Alternative splicing results in multiple transcript variants.
Specifications
| DP2 | |
| Polyclonal | |
| Unconjugated | |
| TFDP2 | |
| 1110029I05Rik; A330080J22Rik; BTPS2; DP2; DP2.5; DP3; DP-3; E2F dimerization partner 2; GS; Tcfdp2; TFDP2; transcription factor Dp 2; transcription factor Dp-2; transcription factor Dp-2 (E2F dimerization partner 2); Unknown (protein for MGC:133813) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 300947 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| TFDP2 | |
| synthetic peptide directed towards the N-terminal region of rat DP2. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction