missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15897003
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15897003 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15897003 Supplier Invitrogen™ Supplier No. PA568596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 100%; Rat: 93%.

The gene is a member of the transcription factor DP family. The encoded protein forms heterodimers with the E2F transcription factors resulting in transcriptional activation of cell cycle regulated genes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DP2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TFDP2
Gene Accession No. Q14188
Gene Alias 1110029I05Rik; A330080J22Rik; BTPS2; DP2; DP2.5; DP3; DP-3; E2F dimerization partner 2; GS; Tcfdp2; TFDP2; transcription factor Dp 2; transcription factor Dp-2; transcription factor Dp-2 (E2F dimerization partner 2); Unknown (protein for MGC:133813)
Gene Symbols TFDP2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human DP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7029
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.