missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31880
This item is not returnable.
View return policy
Description
DP1 Polyclonal specifically detects DP1 in Human samples. It is validated for Western Blot.
Specifications
| DP1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal | |
| TFDP1 | |
| Recombinant protein encompassing a sequence within the center region of human DP1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Chimpanzee (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.68mg/mL | |
| Western Blot 1:500-1:3000 | |
| DP1E2F dimerization partner 1, DRTF1Dp-1, DRTF1-polypeptide 1, E2F-related transcription factor, transcription factor Dp-1 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 7027 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction