missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxylamine Succinate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Doxilamine succinate |
| CAS | 562-10-7 |
| Molecular Formula | C17 H22 N2 O . C4 H6 O4 |
| Quantity | 1 g |
| Synonym | Doxylamine Hydrogen Succinate, Butanedioic acid, compd. with N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]ethanamine (1:1), Succinic acid, compd. with 2-[α-[2-(dimethylamino)ethoxy]-α-methylbenzyl]pyridine (1:1) (8CI), Ethanamine, N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]-, butanedioate (1:1) (9CI), Pyridine, 2-[α-[2-(dimethylamino)ethoxy]-α-methylbenzyl]-, succinate (1:1) (8CI), 2-[α-(2-Dimethylaminoethoxy)-α-methylbenzyl]pyridine succinate, 2-[α-(2-Dimethylaminoethoxy)-α-methylbenzyl]pyridine succinate (1:1), Alsodorm, Decamium, Decapryn, Decapryn succinate, Decapryn succinate (1:1), Donormil, Donormyl, Doxised, Doxylamine succinate (1:1), Gittalun, Hoggar N, Meraprina, Mereprine, NSC 74772, Sedaplus, Unisom |
| InChI Formula | InChI=1S/C17H22N2O.C4H6O4/c1-17(20-14-13-19(2)3,15-9-5-4-6-10-15)16-11-7-8-12-18-16;5-3(6)1-2-4(7)8/h4-12H,13-14H2,1-3H3;1-2H2,(H,5,6)(H,7,8) |
| SMILES | CN(C)CCOC(C)(c1ccccc1)c2ccccn2.OC(=O)CCC(=O)O |
| IUPAC Name | butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine |
| Molecular Weight (g/mol) | 388.46 |
| Formula Weight | 388.1998 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction