missing translation for 'onlineSavingsMsg'
Learn More

Doxylamine succinate, 98%

Product Code. 13535040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13535040 5 g Glass bottle 5g
13545040 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13535040 Supplier Thermo Scientific Acros Supplier No. 449540050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 562-10-7 | C21H28N2O5 | 388.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562-10-7
Molecular Formula C21H28N2O5
Molecular Weight (g/mol) 388.46
MDL Number MFCD00056168
InChI Key KBAUFVUYFNWQFM-UHFFFAOYNA-N
Synonym doxylamine succinate, decapryn, evigoa d, a-h injection, decapryn succinate, unisom, alsadorm, gittalun, mereprine
PubChem CID 11224
ChEBI CHEBI:82461
IUPAC Name butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine
SMILES OC(=O)CCC(O)=O.CN(C)CCOC(C)(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 388.46
Chemical Name or Material Doxylamine succinate
compliance-icons
Product Identifier
  • Doxylamine succinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.