missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline hydrochloride

Product Code. p-7041833
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15473189

Brand: Thermo Scientific Alfa Aesar J60422.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Doxycycline hydrochloride, CAS # 10592-13-9, is the hydrochloride salt form of doxycycline that shows antibacterial and antimalarial activities. | CAS: 10592-13-9 | C22H25ClN2O8 | 480.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

  • Doxycycline hydrochloride is a compound that shows antibacterial and antimicrobial activities against several species, including Pseudomonas aeruginosa, Plasmodium falciparum, and Escherichia coli. It is also a bacteriostatic
  • It can block the binding of aminoacyl-tRNA to the mRNA-ribosome complex, thereby inhibiting protein synthesis

Application

  • It is used to eliminate Borrelia burgdorferi and Anaplasma phagocytophilum in rodent reservoirs and eliminate Ixodes scapularis ticks
  • Doxycycline hydrochloride is also considered to be a broad-spectrum inhibitor used to inhibit matrix metalloproteinases (MMP), such as type 1 collagenase, in wound healing and tissue remodeling studies. Due to this activity, it can inhibit angiogenesis and reduce lung metastases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10592-13-9
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.90
MDL Number MFCD03427564
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
PubChem CID 54706018
IUPAC Name (1S,3Z,4aS,11R,11aR,12S,12aR)-3-[amino(hydroxy)methylidene]-4a,6,7,12-tetrahydroxy-N,N,11-trimethyl-2,4,5-trioxo-1,2,3,4,4a,5,11,11a,12,12a-decahydrotetracen-1-aminium chloride
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12

Specifications

Type Doxycycline Hydrochloride
Physical Form Crystalline Powder
Melting Point 195°C to 201°C
Color Yellow
Quantity 5 g
Solubility Information Soluble in water to 50mg/ml; May require warming.
Formula Weight 480.9
Chemical Name or Material Doxycycline hydrochloride
compliance-icons
Product Identifier
  • Doxycycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Doxycycline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.