missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hyclate, TRC

Product Code. 17960525
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960525 50 mg 50mg
17911875 1 g 1g
17924095 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17960525 Supplier Toronto Research Chemicals Supplier No. TRCD56150050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24390-14-5
Molecular Formula 2 C22 H24 N2 O8 . C2 H6 O . 2 Cl H . H2 O
Molecular Weight (g/mol) 1025.87
Synonym Doxycycline Hyclate, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride, (4S,4aR,5S,5aR,6R,12aS)-, compd. with ethanol, hydrate (2:2:1:1), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, (4S,4aR,5S,5aR,6R,12aS)-, compd. with ethanol (2:1), monohydrate (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5α,5aα,6α,12aα)]-, compd. with ethanol (2:1), monohydrate, 2-Naphthacenecarboxamide, 4β-(dimethylamino)-1,4,4aβ,5,5aβ,6,11,12a-octahydro-3,5β,10,12,12aβ-pentahydroxy-6β-methyl-1,11-dioxo-, monohydrochloride, compd. with ethyl alc. (2:1), monohydrate (8CI), Ethanol, compd. with (4S,4aR,5S,5aR,6R,12aS)-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride (1:2), monohydrate (9CI), Ethanol, compd. with [4S-(4α,4aα,5α,5aα,6α,12aα)]-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride (1:2), monohydrate, 6-Desoxy-5-hydroxytetracycline hydrochloride hemihydrate hemiethanolate, Atridox, BioPure MTAD, DOX-T 100, DOXY 100, Doxitrat, Doxycycline hyclate, Doxycycline hydrochloride hemiethanolate hemihydrate, Doxyprex, Monodoks, Periostat, Tetraclean, WC 2031
IUPAC Name (4S,4aR,5S,5aR,6R,12aS)-4-(dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;hydrate;dihydrochloride
SMILES O.Cl.Cl.CCO.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=C(C(=O)N)C(=O)[C@@]3(O)C(=C2C(=O)c4c(O)cccc14)O)O.C[C@@H]5[C@H]6[C@H](O)[C@H]7[C@H](N(C)C)C(=C(C(=O)N)C(=O)[C@@]7(O)C(=C6C(=O)c8c(O)cccc58)O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);3H,2H2,1H3;2*1H;1H2/t2*7-,10+,14+,15-,17-,22-;;;;/m00..../s1
Formula Weight 1024.3123
Chemical Name or Material Doxycycline Hyclate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.