missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hyclate, Mikromol™

Product Code. 17913806
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17913806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913806 Supplier Mikromol Supplier No. MM0276.00
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Doxycycline Hyclate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Doxycycline Hyclate
CAS 24390-14-5
Analyte or Component Names Doxycycline Hyclate
Shipping Condition Room Temperature
Molecular Formula 2 C22 H24 N2 O8 . C2 H6 O . 2 Cl H . H2 O
Certifications/Compliance ISO 9001
Synonym Doxycycline Hyclate, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, (4S,4aR,5S,5aR,6R,12aS)-, compd. with ethanol (2:1), monohydrate (9CI), Ethanol, compd. with (4S,4aR,5S,5aR,6R,12aS)-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride (1:2), monohydrate (9CI), 6-Desoxy-5-hydroxytetracycline hydrochloride hemihydrate hemiethanolate, Atridox, BioPure MTAD, DOX-T 100, DOXY 100, Doxitrat, Doxycycline hyclate, Doxycycline hydrochloride hemiethanolate hemihydrate, Doxyprex, Monodoks, Periostat, Tetraclean, WC 2031, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, [4S-(4alpha,4aalpha,5alpha,5aalpha,6alpha,12aalpha)]-, compd. with ethanol (2:1), monohydrate, 2-Naphthacenecarboxamide, 4beta-(dimethylamino)-1,4,4abeta,5,5abeta,6,11,12a-octahydro-3,5beta,10,12,12abeta-pentahydroxy-6beta-methyl-1,11-dioxo-, monohydrochloride, compd. with ethyl alc. (2:1), monohydrate (8CI), Ethanol, compd. with [4S-(4alpha,4aalpha,5alpha,5aalpha,6alpha,12aalpha)]-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride (1:2), monohydrate
InChI Formula InChI=1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);3H,2H2,1H3;2*1H;1H2/t2*7-,10+,14+,15-,17-,22-;;;;/m00..../s1
SMILES OC1=C2C([C@H](C)C3C(C2=O)=C(O)[C@@]4(O)C([C@H](N(C)C)C(O)=C(C(N)=O)C4=O)[C@H]3O)=CC=C1.CCO.OC5=C6C([C@H](C)C7C(C6=O)=C(O)[C@@]8(O)C([C@H](N(C)C)C(O)=C(C(N)=O)C8=O)[C@H]7O)=CC=C5.Cl.O.Cl
IUPAC Name (4S,4aR,5S,5aR,6R,12aS)-4-(dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;hydrate;dihydrochloride
Molecular Weight (g/mol) 1025.87
Quantity 100 mg
Formula Weight 1024.3123 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.