missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Doxycycline (Hyclate)

Product Code. 17100122
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17100122 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17100122 Supplier STEMCELL Technologies Supplier No. 72742
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibiotic; Used in Tet-inducible gene expression systems

  • Doxycycline is a tetracycline-like antibiotic (Hicks et al.; Nau et al.; Solís García del Pozo & Solera; Steinhardt et al.) In genetic engineering, doxycycline is used as the regulator for inducible gene expression systems, whereby expression depends on either the presence (Tet-On) or absence (Tet-Off ) of doxycycline
  • Also, doxycycline inhibits certain matrix metalloproteinases (MMP), such as MMP-8 (Ki = 36 μM; Griffin et al.; Smith et al.) It only poorly inhibits MMP-1 and MMP-13 (Ki > 100 μM; Smith et al.) This product is supplied as the hyclate salt of the molecule
  • Used as the regulator for inducible gene expression in lentiviral infection and transgenic mouse models using the Tet-On or Tet-Off systems (Brambrink et al.; Carey et al.; Haenebalcke et al.; Hanna et al.; Hockemeyer et al.; Maherali et al.; Markoulaki et al.; Stadtfeld et al. 2008, 2010; Wernig et al.)

Specifications

CAS 24390-14-5
Synonym WC 2031
Quantity 1 g
Molecular Formula 2[C22H24N2O8] · 2HCl · H2O · C2H6O
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.