missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin hydrochloride, MedChemExpress

Product Code. 16406646
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16406646

Brand: Medchem Express HY15142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxorubicin (Hydroxydaunorubicin) hydrochloride, a cytotoxic anthracycline antibiotic, is an anti-cancer chemotherapy agent.

Doxorubicin hydrochloride is a potent human DNA topoisomerase I and topoisomerase II inhibitor with IC50s of 0.8 μM and 2.67 μM, respectively. Doxorubicin hydrochloride reduces basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. Doxorubicin hydrochloride induces apoptosis and autophagy.

Specifications

Chemical Name or Material Doxorubicin hydrochloride
Physical Form Solid
CAS 25316-40-9
Color Reddish Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C27H30ClNO11
For Use With (Application) Cancer-Kinase/protease
Synonym Hydroxydaunorubicin hydrochloride
Solubility Information DMSO : 35.71 mg/mL (61.57 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 20 mg/mL (34.48 mM; Need ultrasonic)
SMILES COC1=C2C(C(C(C(O)=C(C[C@](C(CO)=O)(O)C[C@@H]3O[C@@]4([H])C[C@H](N)[C@H](O)[C@H](C)O4)C3=C5O)=C5C2=O)=O)=CC=C1.[H]Cl
Molecular Weight (g/mol) 579.98
Formula Weight 579.98
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.