missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin hydrochloride, MedChemExpress

Product Code. 16452437
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452437 50 mg 50mg
16451756 500 mg 500mg
16421166 200 mg 200mg
16406646 100 mg 100mg
16431479 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16452437 Supplier Medchem Express Supplier No. HY15142
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Doxorubicin (Hydroxydaunorubicin) hydrochloride, a cytotoxic anthracycline antibiotic, is an anti-cancer chemotherapy agent.

Doxorubicin hydrochloride is a potent human DNA topoisomerase I and topoisomerase II inhibitor with IC50s of 0.8 μM and 2.67 μM, respectively. Doxorubicin hydrochloride reduces basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. Doxorubicin hydrochloride induces apoptosis and autophagy.

Specifications

Chemical Name or Material Doxorubicin hydrochloride
Physical Form Solid
CAS 25316-40-9
Color Reddish Brown
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H30ClNO11
For Use With (Application) Cancer-Kinase/protease
Synonym Hydroxydaunorubicin hydrochloride
Solubility Information DMSO : 35.71 mg/mL (61.57 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 20 mg/mL (34.48 mM; Need ultrasonic)
SMILES COC1=C2C(C(C(C(O)=C(C[C@](C(CO)=O)(O)C[C@@H]3O[C@@]4([H])C[C@H](N)[C@H](O)[C@H](C)O4)C3=C5O)=C5C2=O)=O)=CC=C1.[H]Cl
Molecular Weight (g/mol) 579.98
Formula Weight 579.98
Percent Purity 95.0%
Grade Research
Show More Show Less
compliance-icons
Product Identifier
  • Doxorubicin (hydrochloride)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P313-Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Hydroxydaunorubicin(hydrochloride)

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.