missing translation for 'onlineSavingsMsg'
Learn More

Doxofylline

Product Code. 15483319
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483319 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15483319 Supplier Thermo Scientific Alfa Aesar Supplier No. J60575.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 69975-86-6 | C11H14N4O4 | 266.257 g/mol

Xanthine bronchodilator that inhibits phosphodiesterase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 69975-86-6
Melting Point 144°C to 146°C
Color White
Molecular Formula C11H14N4O4
MDL Number MFCD00865218
Quantity 5 g
Merck Index 14,3438
Synonym doxofylline, doxophylline, ansimar, dioxyfilline, maxivent, ventax, unii-mpm23gmo7z, doxofylline usan:inn, doxofilina inn-spanish, doxofyllinum inn-latin
InChI Key HWXIGFIVGWUZAO-UHFFFAOYSA-N
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC3OCCO3
IUPAC Name 7-(1,3-dioxolan-2-ylmethyl)-1,3-dimethylpurine-2,6-dione
Molecular Weight (g/mol) 266.257
PubChem CID 50942
Formula Weight 266.25
Physical Form Crystalline Powder
Chemical Name or Material Doxofylline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.