missing translation for 'onlineSavingsMsg'
Learn More

Doxifluridine, MedChemExpress

Product Code. 16470758
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16470758 5 g 5g
16412039 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16470758

Brand: Medchem Express HYB0021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxifluridine(Ro 21-9738; 5'-DFUR) is a thymidine phosphorylase activator for PC9-DPE2 cells with IC50 of 0.62 μM.

Doxifluridine(Ro 21-9738

Specifications

Chemical Name or Material Doxifluridine
CAS 9-5-3094
Color White
Purity Grade Notes Research
Molecular Formula C9H11FN2O5
Quantity 5 g
Synonym Ro 21-9738 5-Fluoro-5'-deoxyuridine 5'-DFUR
Solubility Information DMSO : ≥ 100 mg/mL (406.19 mM) ∣DMF : 100 mg/mL (406.19 mM; Need ultrasonic) ∣H2O : 20 mg/mL (81.24 mM; Need ultrasonic)
SMILES C[C@@H]1[C@H]([C@H]([C@H](N2C(NC(C(F)=C2)=O)=O)O1)O)O
Molecular Weight (g/mol) 246.19
Formula Weight 246.19
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.