missing translation for 'onlineSavingsMsg'
Learn More

Doxifluridine, MedChemExpress

Product Code. 16412039
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16412039

Brand: Medchem Express HYB0021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxifluridine(Ro 21-9738; 5'-DFUR) is a thymidine phosphorylase activator for PC9-DPE2 cells with IC50 of 0.62 μM.

Doxifluridine(Ro 21-9738

Specifications

Chemical Name or Material Doxifluridine
CAS 9-5-3094
Color White
Purity Grade Notes Research
Molecular Formula C9H11FN2O5
Quantity 1 g
Synonym Ro 21-9738 5-Fluoro-5'-deoxyuridine 5'-DFUR
Solubility Information DMSO : ≥ 100 mg/mL (406.19 mM) ∣DMF : 100 mg/mL (406.19 mM; Need ultrasonic) ∣H2O : 20 mg/mL (81.24 mM; Need ultrasonic)
SMILES C[C@@H]1[C@H]([C@H]([C@H](N2C(NC(C(F)=C2)=O)=O)O1)O)O
Molecular Weight (g/mol) 246.19
Formula Weight 246.19
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.