missing translation for 'onlineSavingsMsg'
Learn More

Doxepin hydrochloride

Product Code. 15414929
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414929 25 g 25g
15404929 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414929 Supplier Thermo Scientific Alfa Aesar Supplier No. J62579.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent H1-histamine receptor antagonist and a 5-HT receptor antagonist | CAS: 1229-29-4 | C19H22ClNO | 315.841 g/mol

It is a highly potent H1 histamine receptor antagonist (Kd = 310 pM) and tricyclic antidepressant that inhibits the reuptake of serotonin and norepinephrine and acts as an antagonist at histamine, serotonin, adrenergic, and muscarinic receptors with Ki values in the nanomolar range. Also binds to the H4 histamine receptor (pKi = 6.79). Used clinically to treat anxiety and depression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a highly potent H1 histamine receptor antagonist (Kd = 310 pM) and tricyclic antidepressant that inhibits the reuptake of serotonin and norepinephrine and acts as an antagonist at histamine, serotonin, adrenergic, and muscarinic receptors with Ki values in the nanomolar range. Also binds to the H4 histamine receptor (pKi = 6.79). Used clinically to treat anxiety and depression.

Solubility
Soluble in water (100 mM), chloroform (1:2), alcohol (1:1), methanol, and DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1229-29-4
Molecular Formula C19H22ClNO
Molecular Weight (g/mol) 315.841
MDL Number MFCD00079135
InChI Key MHNSPTUQQIYJOT-SJDTYFKWSA-N
Synonym doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride
PubChem CID 6419921
IUPAC Name (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl

Specifications

Melting Point 187°C to 189°C
Color White
Quantity 25 g
UN Number UN2811
Merck Index 14,3435
Solubility Information Soluble in water (100 mM),chloroform (1:2),alcohol (1:1),methanol,and DMSO.
Formula Weight 315.84
Physical Form Powder
Chemical Name or Material Doxepin hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.