missing translation for 'onlineSavingsMsg'
Learn More

Doxepin Hydrochloride, MedChemExpress

Product Code. p-200099114
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 16468588

Brand: Medchem Express HYB0725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxepin hydrochloride is an orally active tricyclic antidepressant agent.

Doxepin hydrochloride is a potent and selective histamine receptor H1 antagonist. Doxepin hydrochloride is also a potent CYP450 inhibitor and significantly inhibits CYP450 2C19 and 1A2. Doxepin inhibits reuptake of serotonin and norepinephrine as a tricyclic antidepressant.. Doxepin has therapeutic effects in atopic dermatitis,chronic urticarial,can improve cognitive processes, protect central nervous system.. Doxepin has also been proposed as a protective factor against oxidative stress..

Specifications

Chemical Name or Material Doxepin Hydrochloride
CAS 1229-29-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H22ClNO
Quantity 10 g
Solubility Information DMSO : ≥ 100 mg/mL (316.62 mM) ∣H2O : ≥ 50 mg/mL (158.31 mM)
SMILES CN(C)CC/C=C1C2=CC=CC=C2OCC3=CC=CC=C/13.Cl
Molecular Weight (g/mol) 315.84
Formula Weight 315.84
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.