missing translation for 'onlineSavingsMsg'
Learn More

Doxepin.HCl, Lipomed

Product Code. 30461812
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461812 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461812 Supplier Lipomed Supplier No. LPMDXP1092HC50
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Doxepin.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Doxepin Hydrochloride
CAS 1229-29-4
Analyte or Component Names Doxepin Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H21 N O . Cl H
Synonym Doxepin Hydrochloride, 3-Dibenz[b,e]oxepin-11(6 H)-ylidene-N,N-dimethyl-1-propanamine hydrochloride, Dibenz[b,e]oxepin, 1-propanamine deriv., Adapin, Aponal, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6 H),gamma-propylamine hydrochloride
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CN(C)CC\C=C\1/c2ccccc2COc3ccccc13
IUPAC Name (3 E)-3-(6 H-benzo[c][1 ]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 315.84
Quantity 50 mg
Formula Weight 315.139 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.