missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxazosin Mesylate, TRC
Specifications
Specifications
| Chemical Name or Material | Doxazosin mesilate |
| CAS | 77883-43-3 |
| Molecular Formula | C23 H25 N5 O5 . C H4 O3 S |
| Quantity | 1 g |
| Synonym | Doxazosin Mesilate, Methanone, [4-(4-amino-6,7-dimethoxy-2-quinazolinyl)-1-piperazinyl](2,3-dihydro-1,4-benzodioxin-2-yl)-, methanesulfonate (1:1), Piperazine, 1-(4-amino-6,7-dimethoxy-2-quinazolinyl)-4-[(2,3-dihydro-1,4-benzodioxin-2-yl)carbonyl]-, monomethanesulfonate (9CI), 1,4-Benzodioxin, piperazine deriv., Alfadil, Cardenalin, Cardran, Cardular, Cardura, Cardura (pharmaceutical), Carduran, Diblocin, Doxazosin mesylate, Doxazosin monomethanesulfonate, Normothen, Supressin, UK 33274-27, doxazolazine mesylate |
| InChI Formula | InChI=1S/C23H25N5O5.CH4O3S/c1-30-18-11-14-15(12-19(18)31-2)25-23(26-21(14)24)28-9-7-27(8-10-28)22(29)20-13-32-16-5-3-4-6-17(16)33-20;1-5(2,3)4/h3-6,11-12,20H,7-10,13H2,1-2H3,(H2,24,25,26);1H3,(H,2,3,4) |
| SMILES | COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4COc5ccccc5O4.CS(=O)(=O)O |
| IUPAC Name | [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(2,3-dihydro-1,4-benzodioxin-3-yl)methanone;methanesulfonic acid |
| Molecular Weight (g/mol) | 547.58 |
| Formula Weight | 547.1737 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?