missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxazosin mesylate, MedChemExpress
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Description
Doxazosin mesylate (UK 33274) is a quinazoline-derivative that selectively antagonizes postsynaptic α1-adrenergic receptors.
Specifications
Specifications
| Chemical Name or Material | Doxazosin mesylate |
| CAS | 77883-43-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H29N5O8S |
| Quantity | 500 mg |
| Synonym | UK 33274 mesylate |
| Solubility Information | DMSO : 33.33 mg/mL (60.87 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.83 mM; Need ultrasonic) |
| SMILES | O=C(N1CCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)CC1)C4OC5=CC=CC=C5OC4.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 547.58 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction