missing translation for 'onlineSavingsMsg'
Learn More

Doxazosin EP Impurity D (Doxazosin USP Related Compound B), Mikromol™

Product Code. 30049362
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049362 25 mg 25mg
17903006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049362 Supplier Mikromol Supplier No. MM0085.070025
 Restricted Item
Estimated Shipment: 23-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Doxazosin EP Impurity D (Doxazosin USP Related Compound B)
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 6,7-dimethoxyquinazoline-2,4(1H,3H)-dione
CAS 28888-44-0
Analyte or Component Names 6,7-dimethoxyquinazoline-2,4(1H,3H)-dione
Shipping Condition Room Temperature
Molecular Formula C10 H10 N2 O4
Certifications/Compliance ISO 17025
Synonym 6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione, Doxazosin Mesilate Imp. D (EP), Doxazosin USP Related Compound B, Doxazosin USP RC B, 6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione, 2,4(1H,3H)-Quinazolinedione, 6,7-dimethoxy-, 6,7-Dimethoxy-2,4(1H,3H)-quinazolinedione, 2,5-Dihydroxy-6,7-dimethoxyquinazoline, 6,7-Dimethoxy-1H-quinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-diol, 6,7-Dimethoxyquinazoline-2,4-dione, Doxazosin Related Compound B, Doxazosin Mesilate Impurity D, Doxazosin Impurity D
InChI Formula InChI=1S/C10H10N2O4/c1-15-7-3-5-6(4-8(7)16-2)11-10(14)12-9(5)13/h3-4H,1-2H3,(H2,11,12,13,14)
SMILES COc1cc2NC(=O)NC(=O)c2cc1OC
IUPAC Name 6,7-dimethoxy-1H-quinazoline-2,4-dione
Molecular Weight (g/mol) 222.2
Quantity 25 mg
Formula Weight 222.0641 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.