missing translation for 'onlineSavingsMsg'
Learn More

Doxapram Hydrochloride Monohydrate, Mikromol™

Product Code. 18083080
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083080 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18083080 Supplier Mikromol Supplier No. MM0525.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Doxapram Hydrochloride Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Doxapram hydrochloride hydrate
CAS 7081-53-0
Analyte or Component Names Doxapram hydrochloride hydrate
Shipping Condition Room Temperature
Molecular Formula C24 H30 N2 O2 . Cl H . H2 O
Certifications/Compliance ISO 17025
Synonym Doxapram hydrochloride, (4RS)-1-Ethyl-4-[2-(morpholin-4-yl)ethyl]-3,3-diphenylpyrrolidin-2-one hydrochloride, 2-Pyrrolidinone, 1-ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-, hydrochloride, hydrate (1:1:1), 2-Pyrrolidinone, 1-ethyl-4-(2-morpholinoethyl)-3,3-diphenyl-, monohydrochloride, monohydrate (8CI), 2-Pyrrolidinone, 1-ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-, monohydrochloride, monohydrate (9CI), 1-Ethyl-4-(2-morpholinoethyl)-3,3-diphenyl-2-pyrrolidinone hydrochloride hydrate, AHR 619, Doxapram hydrochloride hydrate, Doxapram hydrochloride monohydrate
InChI Formula InChI=1S/C24H30N2O2.ClH.H2O/c1-2-26-19-22(13-14-25-15-17-28-18-16-25)24(23(26)27,20-9-5-3-6-10-20)21-11-7-4-8-12-21;;/h3-12,22H,2,13-19H2,1H3;1H;1H2
SMILES O.Cl.CCN1CC(CCN2CCOCC2)C(C1=O)(c3ccccc3)c4ccccc4
IUPAC Name 1-ethyl-4-(2-morpholin-4-ylethyl)-3,3-diphenylpyrrolidin-2-one;hydrate;hydrochloride
Molecular Weight (g/mol) 432.98
Quantity 250 mg
Formula Weight 432.218 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.