missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxapram Hydrochloride Monohydrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Doxapram hydrochloride hydrate |
| CAS | 7081-53-0 |
| Analyte or Component Names | Doxapram hydrochloride hydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H30 N2 O2 . Cl H . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Doxapram hydrochloride, (4RS)-1-Ethyl-4-[2-(morpholin-4-yl)ethyl]-3,3-diphenylpyrrolidin-2-one hydrochloride, 2-Pyrrolidinone, 1-ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-, hydrochloride, hydrate (1:1:1), 2-Pyrrolidinone, 1-ethyl-4-(2-morpholinoethyl)-3,3-diphenyl-, monohydrochloride, monohydrate (8CI), 2-Pyrrolidinone, 1-ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-, monohydrochloride, monohydrate (9CI), 1-Ethyl-4-(2-morpholinoethyl)-3,3-diphenyl-2-pyrrolidinone hydrochloride hydrate, AHR 619, Doxapram hydrochloride hydrate, Doxapram hydrochloride monohydrate |
| InChI Formula | InChI=1S/C24H30N2O2.ClH.H2O/c1-2-26-19-22(13-14-25-15-17-28-18-16-25)24(23(26)27,20-9-5-3-6-10-20)21-11-7-4-8-12-21;;/h3-12,22H,2,13-19H2,1H3;1H;1H2 |
| Solution Type | Neat |
| SMILES | O.Cl.CCN1CC(CCN2CCOCC2)C(C1=O)(c3ccccc3)c4ccccc4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?