missing translation for 'onlineSavingsMsg'
Learn More

Doxapram Hydrochloride Hydrate, TRC

Product Code. 30384683
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384683 10 mg
30385219 50 mg
30375063 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384683 Supplier Toronto Research Chemicals Supplier No. TRCD55250010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7081-53-0
Molecular Formula C24 H30 N2 O2 . Cl H . H2 O
Molecular Weight (g/mol) 432.98
Synonym Doxapram hydrochloride, (4RS)-1-Ethyl-4-[2-(morpholin-4-yl)ethyl]-3,3-diphenylpyrrolidin-2-one hydrochloride, 2-Pyrrolidinone, 1-ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-, hydrochloride, hydrate (1:1:1), 2-Pyrrolidinone, 1-ethyl-4-(2-morpholinoethyl)-3,3-diphenyl-, monohydrochloride, monohydrate (8CI), 2-Pyrrolidinone, 1-ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-, monohydrochloride, monohydrate (9CI), 1-Ethyl-4-(2-morpholinoethyl)-3,3-diphenyl-2-pyrrolidinone hydrochloride hydrate, AHR 619, Doxapram hydrochloride hydrate, Doxapram hydrochloride monohydrate
IUPAC Name 1-ethyl-4-(2-morpholin-4-ylethyl)-3,3-diphenylpyrrolidin-2-one;hydrate;hydrochloride
SMILES O.Cl.CCN1CC(CCN2CCOCC2)C(C1=O)(c3ccccc3)c4ccccc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H30N2O2.ClH.H2O/c1-2-26-19-22(13-14-25-15-17-28-18-16-25)24(23(26)27,20-9-5-3-6-10-20)21-11-7-4-8-12-21;;/h3-12,22H,2,13-19H2,1H3;1H;1H2
Formula Weight 432.218
Chemical Name or Material Doxapram hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.