missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Chemical Name or Material | Doxapram hydrochloride hydrate |
| CAS | 7081-53-0 |
| Quantity | 250 mg |
| Solubility | Soluble aqueous solution: slightly soluble in chloroform |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C24 H30 N2 O2 . H Cl . x H2 O |
| Physical Form | Crystalline Powder |
| Color | White to Almost-white |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?