missing translation for 'onlineSavingsMsg'
Learn More

Doxapram hydrochloride hydrate, MedChemExpress

Product Code. 16468366
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468366 100 mg 100mg
16434397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468366 Supplier Medchem Express Supplier No. HYB0551A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxapram hydrochloride hydrate inhibits TASK-1, TASK-3, TASK-1/TASK-3 heterodimeric channel function with EC50 of 410 nM, 37 μM, 9 μM, respectively.

Doxapram hydrochloride hydrate inhibits TASK-1, TASK-3, TASK-1/TASK-3 heterodimeric channel function with EC50 of 410 nM, 37 μM, 9 μM, respectively.

Specifications

Chemical Name or Material Doxapram hydrochloride hydrate
CAS 7081-53-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H33ClN2O3
Quantity 100 mg
Solubility Information DMSO : ≥ 55 mg/mL (127.03 mM) ∣H2O : 25 mg/mL (57.74 mM; Need ultrasonic)
SMILES O=C1N(CC)CC(CCN2CCOCC2)C1(C3=CC=CC=C3)C4=CC=CC=C4.Cl.O
Molecular Weight (g/mol) 432.98
Formula Weight 432.98
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.