missing translation for 'onlineSavingsMsg'
Learn More

Doxapram hydrochloride hydrate, MedChemExpress

Product Code. 16434397
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16434397

Brand: Medchem Express HYB0551A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxapram hydrochloride hydrate inhibits TASK-1, TASK-3, TASK-1/TASK-3 heterodimeric channel function with EC50 of 410 nM, 37 μM, 9 μM, respectively.

Doxapram hydrochloride hydrate inhibits TASK-1, TASK-3, TASK-1/TASK-3 heterodimeric channel function with EC50 of 410 nM, 37 μM, 9 μM, respectively.

Specifications

Chemical Name or Material Doxapram hydrochloride hydrate
CAS 7081-53-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H33ClN2O3
Quantity 50 mg
Solubility Information DMSO : ≥ 55 mg/mL (127.03 mM) ∣H2O : 25 mg/mL (57.74 mM; Need ultrasonic)
SMILES O=C1N(CC)CC(CCN2CCOCC2)C1(C3=CC=CC=C3)C4=CC=CC=C4.Cl.O
Molecular Weight (g/mol) 432.98
Formula Weight 432.98
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.