missing translation for 'onlineSavingsMsg'
Learn More

Dowex 1X8 chloride form 100-200 Mesh, TRC

Product Code. 30380680
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380680 1 kg 1kg
30373157 500 g 500g
30371533 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380680 Supplier Toronto Research Chemicals Supplier No. TRCD4855151KG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dowex 1X4 Chloride
CAS 69011-19-4
Molecular Formula (C10 H12 . C10 H10 . C8 H8 . C3 H9 N)x
Quantity 1 kg
Synonym Dowex™ 1X4-200 Ion-Exchange Resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.