missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dowex 1X8 chloride form 100-200 Mesh, TRC
Specifications
Specifications
| Chemical Name or Material | Dowex 1X4 Chloride |
| CAS | 69011-19-4 |
| Molecular Formula | (C10 H12 . C10 H10 . C8 H8 . C3 H9 N)x |
| Quantity | 500 g |
| Synonym | Dowex™ 1X4-200 Ion-Exchange Resin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?