missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™ 1X8 200-400 (Cl)

Product Code. 11406820
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406820 1000 g 1000g
11416820 50 g 50g
11426820 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406820 Supplier Thermo Scientific Alfa Aesar Supplier No. L14258.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol

AmberChrom 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12627-85-9
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132718
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
SMILES *

Specifications

Odor Amine-like
Quantity 1000 g
Solubility Information Insoluble in water.
IUPAC Name 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
Chemical Name or Material AmberChrom 1X8 200-400 (Cl)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.