missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™ 1X8 100-200 (Cl)

Product Code. 11349075
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349075 50 g 50g
11359075 250 g 250g
11369075 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349075 Supplier Thermo Scientific Alfa Aesar Supplier No. L14257.18
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol

AmberChrom™ 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
AmberChrom 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12627-85-9
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132718
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
SMILES *

Specifications

Odor Amine-like
Quantity 50 g
Solubility Information Insoluble in water.
Chemical Name or Material AmberChrom 1X8 100-200 (Cl)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.