missing translation for 'onlineSavingsMsg'
Learn More

DOV-216,303 Free Base, MedChemExpress

Product Code. 16439987
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439987 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439987 Supplier Medchem Express Supplier No. HY18332C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DOV-216,303 (Free Base) is a potent triple serotonin, norepinephrine, and dopamine reuptake inhibitor, with IC50 values of 14 nM, 20 nM and 78 nM for hSERT, hNET and hDAT, respectively. Has antidepressant-like effects and increases monoamine release in the prefrontal cortex of olfactory bulbectomized (OBX) rats.

DOV-216,303 (Free Base) is a potent triple serotonin, norepinephrine, and dopamine reuptake inhibitor, with IC50 values of 14 nM, 20 nM and 78 nM for hSERT, hNET and hDAT, respectively. Has antidepressant-like effects and increases monoamine release in the prefrontal cortex of olfactory bulbectomized (OBX) rats.

Specifications

Chemical Name or Material DOV-216,303 Free Base
CAS 66504-40-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C11H11Cl2N
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (547.96 mM) ∣Ethanol : 100 mg/mL (438.37 mM; Need ultrasonic)
SMILES ClC1=C(Cl)C=C(C=C1)C12CC1CNC2
Molecular Weight (g/mol) 228.12
Formula Weight 228.12
Percent Purity 98.77%
Grade Research
Physical Form Oil
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.