missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Doublecortin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517428
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.
Specifications
| Doublecortin | |
| Polyclonal | |
| Unconjugated | |
| DCX | |
| DBCN; Dbct; DC; Dcn; DCX; doublecortex; doublecortin; doublin; lissencephalin-X; lissencephaly, X-linked (doublecortin); LISX; Lis-X; LIS-XD; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316; SCLH; XLIS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13193, 1641, 84394 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 165 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O43602, O88809, Q9ESI7 | |
| DCX | |
| Synthetic peptide corresponding to human doublecortin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction