missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doublecortin Antibody (OTI4A3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70558G
This item is not returnable.
View return policy
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Doublecortin | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DCX | |
| Full length recombinant protein of human Doublecortin (NP_835365) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4A3 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin) | |
| Mouse | |
| Affinity Purified | |
| Neuroscience, Phospho Specific, Signal Transduction | |
| 1641.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction