Learn More
Ohaus™ Double Tier Braces
Product Code. 15732469 Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
Platform
Unit Size:
Pack of 4
Product Code. 15732469
Brand: Ohaus™ 30400051
Only null left
Add to basket
Add to basket
OHAUS™ Double Tier Braces provides extra support to tiered platforms.
  • For use with platforms, which can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
TRUSTED_SUSTAINABILITY
Specifications
Product Type Braces
Material Stainless Steel
Description Double Tier Braces, Set of 4
For Use With (Equipment) Platform
Packaging Quantity 4/Pk.