Learn More
Fisherbrand™ PTFE Magnetic Stir Bar, Double-sided crosshead
Designed for use with tube-like containers yet very effective as general stirrers. The PTFE-coated magnet stirrers include a crosshead on both sides of the stirrer for greater turbulence.
Brand:  Fisherbrand™ 001.1117
Product Code. 10416982
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Stable, general-purpose magnetic stirrer
  • Designed for use with tube-like containers yet very effective as general stirrers
  • Double-sided crossheads provider greater turbulence during mixing
  • Autoclavable