missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Double Elastic Polypropylene Clip Caps in Wall Dispenser
GREENER_CHOICE

Product Code. p-7227146 Shop All Medicom Products
Click to view available options
Color:
Black
Blue
Green
White
Unit Size:
Pack of 250

Product Code. 17780129

Brand: Kolmi Hopen™ 70.111

Only null left
Add to basket
Add to basket

These double elastic clip caps come in wall-mounted dispensers for convenient use.

  • Covers the full head, as required by the HACCP process
  • Extra comfort with double elastic to ensure excellent retention
  • Ability to use color code for tracking
  • 12 g/m2 fabric weight

Specifications

Material Polypropylene
Color White
Disposable Single-use
Design Type Clip Cap
Sterility Non-sterile
Certifications/Compliance EN ISO 9001
Quantity 250 Pk.
Dimensions (L x W) 40 x 55 cm
Includes Caps and Wall Dispenser
For Use With (Application) Industrial, Laboratory
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.