Learn More
Juchheim Laborgeraete™ Double Cross Bosshead
Click to view available options
Height (Metric):
30 mm
Unit Size:
Each
Description
- High quality and durable design malleable cast iron with powder coating
- For tripod rods from 6 to 16 mm diameter
- Includes safety screws for mounting by hand without twisting or slipping on the tripod rod
- Made in Germany
Specifications
Specifications
| Material | Stainless Steel |
| Height (Metric) | 30 mm |