Learn More
Juchheim Laborgeraete™ Double Bosshead
High quality double socket made of malleable cast iron.
Brand:  Juchheim Laborgeraete™ 04810
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 15818122
  • £10.35 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Durable due to robust malleable cast iron
  • Option of safety screws for fixing by hand or lobe screws for mounting by hand
  • Made in Germany
Specifications
Specifications
Tempered Cast Iron
30 mm
Lobe screws for mounting by hand