Learn More
Kartell™ Double Ball with Net
Product Code. 10706352
Change view
Click to view available options
Diameter (Metric) Outer:
46 mm
53 mm
Unit Size:
Each
Product Code. Diameter (Metric) Outer unitSize
10706352 53 mm Each
10594621 46 mm Each
2 options
Product Code. 10706352
Brand: Kartell™ 0391300
Only null left
Add to basket
Add to basket
Suitable for automatic burettes and flashchromatography columns
Specifications
Material Red rubber
Diameter (Metric) Outer 53 mm