Learn More
Dotriacontane, 98%
Description
Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.
Solubility
Soluble in hot toluene, diethyl ether, benzene and ethanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Tekniske data
Tekniske data
| Melting Point | 68°C to 71°C |
| Density | 0.812 |
| Boiling Point | 300°C to 310°C |
| Quantity | 5 g |
| Beilstein | 1783260 |
| Solubility Information | Soluble in hot toluene,diethyl ether,benzene and ethanol. Insoluble in water. |
| Formula Weight | 450.88 |
| Percent Purity | 98% |
| Chemical Name or Material | Dotriacontane |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.