missing translation for 'onlineSavingsMsg'
Learn More

Dotriacontane, 97%

Product Code. 10145203 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10145203 100 g Plastic bottle 100g
10230743 5 g Glass bottle 5g
10162190 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10145203 Supplier Thermo Scientific Acros Supplier No. 117691000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 544-85-4 | C32H66 | 450.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-85-4
Molecular Formula C32H66
Molecular Weight (g/mol) 450.88
MDL Number MFCD00009411
InChI Key QHMGJGNTMQDRQA-UHFFFAOYSA-N
Synonym n-dotriacontane, bicetyl, unii-7ksv90rn23, dicetyl, lacceran, dotriacontane, n, dotriacontane, acmc-1akq8, 4-01-00-00595 beilstein handbook reference, wln: 32h
PubChem CID 11008
ChEBI CHEBI:36020
IUPAC Name dotriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 67°C to 72°C
Color White to Gray
Boiling Point 467°C
Flash Point 247°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula CH3(CH2)30CH3
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in ethyl ether,very soluble in benzene
Formula Weight 450.86
Percent Purity 97%
Physical Form Shiny Flakes
Chemical Name or Material Dotriacontane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.