missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOT5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144420
This item is not returnable.
View return policy
Description
DOT5 Polyclonal Antibody for Western Blot, ELISA
Thiol-specific peroxidase that catalyzes the reduction of hydrogen peroxide and organic hydroperoxides to water and alcohols, respectively. Plays a role in cell protection against oxidative stress by detoxifying peroxides and as sensor of hydrogen peroxide-mediated signaling events. Has a role in telomere silencing, which is the repression of chromatin structure which leads to a stop in the transcription of nearby genes. Also has a role in the regulation of telomere length. Acts as an alkyl-hydroperoxide reductase in the nucleus during post-diauxic growth. Preferentially reduces alkyl-hydroperoxides rather than hydrogen peroxide. Acts as an antioxidant necessary for stationary phase survival.
Specifications
| DOT5 | |
| Polyclonal | |
| Unconjugated | |
| DOT5 | |
| Peroxiredoxin DOT5 DOT5 YIL010W | |
| Rabbit | |
| Protein G | |
| RUO | |
| 854805 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.9 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P40553 | |
| DOT5 | |
| Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) DOT5 protein (37-118aa). | |
| 100 μL | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction